Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) fluoride tetrahydrate, Puratronic™, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Cobalt(II) fluoride tetrahydrate
CAS 13817-37-3
Purity Grade Puratronic™
Formula Weight 168.99 (96.93 Anhydrous)
MDL Number MFCD00149656
UN Number UN2923
Merck Index 14,2440
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1069306

 
alfa aesar™
1069306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1069314

 
alfa aesar™
1069314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.99 (96.93 Anhydrous)
Chemical Name or Material Cobalt(II) fluoride tetrahydrate
TSCA TSCA
Odor Odorless
Purity Grade Puratronic™
Physical Form Powder
Assay 99.99%
Sensitivity Moisture Sensitive
Assay Percent Notes (metals basis)
MDL Number MFCD00149656
CAS 13817-37-3
UN Number UN2923
Merck Index 14,2440
Color Pink
Melting Point 200°C (dec.)
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318-H351-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 2.19g/mL
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318-H351-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.