missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) fluoride tetrahydrate, Puratronic™, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1069314

 View more versions of this product

Catalog No. AA1069314



Specifications

Specifications

Cobalt(II) fluoride tetrahydrate
(metals basis)
MFCD00149656
14,2440
MNHUTFNEBYCZAF-UHFFFAOYSA-L
cobalt(2+);difluoride;tetrahydrate
24884271
Powder
Puratronic™
Odorless
Pink
25g
13817-37-3
CoF2H8O4
UN2923
acmc-1btz4, cobalt 2+ fluoride-water 1/2/4, cobalt 2+ tetrahydrate difluoride, cobalt fluoride cof2 ,tetrahydrate 8ci,9ci, cobalt ii ; fluoride tetrahydrate, cobaltous fluoride 4h2o
O.O.O.O.[F-].[F-].[Co+2]
168.99
168.99 (96.93 Anhydrous)
99.99%
2.19 g/mL
Moisture sensitive
200°C (decomposition)

Chemical Identifiers

13817-37-3
168.99
MNHUTFNEBYCZAF-UHFFFAOYSA-L
24884271
O.O.O.O.[F-].[F-].[Co+2]
CoF2H8O4
MFCD00149656
acmc-1btz4, cobalt 2+ fluoride-water 1/2/4, cobalt 2+ tetrahydrate difluoride, cobalt fluoride cof2 ,tetrahydrate 8ci,9ci, cobalt ii ; fluoride tetrahydrate, cobaltous fluoride 4h2o
cobalt(2+);difluoride;tetrahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H351-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P281-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H314-H334-H335-H350

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

EINECSNumber : 233-061-9

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents