Please login to your online account to display your discounted pricing

Cobalt(II) fluoride, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Cobalt(II) fluoride
Name Note 99%
CAS 10026-17-2
Assay ≥98.5% (ex F)
Assay Percent Range 99%
Molecular Formula CoF2
Linear Formula CoF
Formula Weight 96.96
MDL Number MFCD00010941
Synonym Cobalt difluoride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC311270250

  SDS
thermo fisher scientific
311270250
Plastic bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5% (ex F)
Physical Form Powder
Formula Weight 96.96
Quantity 25g
Color Pink
Merck Index 15, 2424
EINECS Number 233-061-9
TSCA TSCA
RTECS Number GG0770000
Packaging Plastic bottle
Assay Percent Range 99%
Molecular Formula CoF2
Linear Formula CoF
MDL Number MFCD00010941
Synonym Cobalt difluoride
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Chemical Name or Material Cobalt(II) fluoride
Name Note 99%
CAS 10026-17-2
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.