Learn More
Cobalt(II) chromate, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 3920309
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cobalt(II) chromate | |
13455-25-9 | |
10 g | |
MFCD00053962 | |
cobalt ii chromate, cobalt 2+ ; dioxido dioxo chromium, cobaltous chromate iii, cobalt chromate ous, cobaltous chromate, acmc-1c0r0, cobalt 2+ chromate 2-, chromic acid h2cro4 , cobalt 2+ salt 1:1 | |
XTUHPOUJWWTMNC-UHFFFAOYSA-N | |
cobalt(2+);dioxido(dioxo)chromium | |
61614 | |
Odorless |
(decomposition) | |
Lump | |
CoCrO4 | |
UN3087 | |
Soluble in alkali and ammomium hydroxide | |
[Co++].[O-][Cr]([O-])(=O)=O | |
174.93 | |
174.93 |
Chemical Identifiers
13455-25-9 | |
174.93 | |
XTUHPOUJWWTMNC-UHFFFAOYSA-N | |
61614 | |
[Co++].[O-][Cr]([O-])(=O)=O |
CoCrO4 | |
MFCD00053962 | |
cobalt ii chromate, cobalt 2+ ; dioxido dioxo chromium, cobaltous chromate iii, cobalt chromate ous, cobaltous chromate, acmc-1c0r0, cobalt 2+ chromate 2-, chromic acid h2cro4 , cobalt 2+ salt 1:1 | |
cobalt(2+);dioxido(dioxo)chromium |
Safety and Handling
GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
P201-P202-P210-P220-P221-P260-P264b-P271-P272-P280g-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c
H272-H314-H317-H334-H335-H350i
DOTInformation : Hazard Class: 5.1; Packaging Group: II
EINECSNumber : 236-651-4
TSCA : Yes
Recommended Storage : Ambient temperatures