missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt(II) chloride, ultra dry, 99.998% (metals basis), Alfa Aesar™

Hygroscopic; 3.367. Decomposes at 400°C on long heating in air

Manufacturer:  Alfa Aesar 1366503

 View more versions of this product

Catalog No. AA1366503



Specifications

Specifications

Cobalt(II) chloride
7646-79-9
Cl2Co
Light blue powder
129.84
3.367g/cm3
Blue
Ultra dry
99.998% (Metals basis)
MFCD00010938
cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii chloride, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride
Powder
Odorless
1g

Chemical Identifiers

7646-79-9
Cl2Co
MFCD00010938
cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii chloride, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride
Safety and Handling

Safety and Handling

H334-H350-H360-H317-H341-H400-H410-H302; May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. May cause an allergic skin reaction. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 231-589-4

SDS Documents