Please login to your online account to display your discounted pricing

Cobalt(II) chloride, ultra dry, 99.998% (metals basis), Alfa Aesar™

Hygroscopic; 3.367. Decomposes at 400°C on long heating in air

Chemical Identifiers

Chemical Name or Material Cobalt(II) chloride
Name Note Ultra dry
CAS 7646-79-9
Physical Form Powder
Assay Percent Range 99.998% (Metals basis)
Molecular Formula Cl2Co
Formula Weight 129.84
MDL Number MFCD00010938
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1366503

  SDS
alfa aesar
1366503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1366506

  SDS
alfa aesar
1366506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1366514

  SDS
alfa aesar
1366514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 129.84
Quantity 1g
Color Light blue
DOT Information DOT Class: 9, Packaging Group: III
Chemical Name or Material Cobalt(II) chloride
Physical Form Powder
EINECS Number 231-589-4
Assay Percent Range 99.998% (Metals basis)
Density 3.367g/cm3
CAS 7646-79-9
MDL Number MFCD00010938
Molecular Formula Cl2Co
Name Note Ultra dry
Appearance Light blue powder
Odor Odorless
Health Hazard 1 H334-H350-H360-H317-H341-H400-H410-H302; May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. May cause an allergic skin reaction. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed.
Safety and Handling

H334-H350-H360-H317-H341-H400-H410-H302; May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. May cause an allergic skin reaction. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed.