Please login to your online account to display your discounted pricing

Cobalt(II) chloride, ultra dry, 99.9% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Cobalt(II) chloride
CAS 7646-79-9
Name Note ultra dry
Molecular Formula Cl2Co
Linear Formula CoCl2
Formula Weight 129.84
MDL Number MFCD00010938
UN Number UN3077
Merck Index 14,2437
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1366603

  SDS
alfa aesar
1366603
Ampouled under Argon 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 129.84
Quantity 1g
Chemical Name or Material Cobalt(II) chloride
TSCA TSCA
Mesh Size -10 Mesh
Odor Odorless
Name Note ultra dry
Packaging Ampouled under Argon
Physical Form Beads
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
Molecular Formula Cl2Co
Linear Formula CoCl2
MDL Number MFCD00010938
CAS 7646-79-9
UN Number UN3077
Merck Index 14,2437
Boiling Point 1049°C
Color Pale Blue
Melting Point 735°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H350-H360-H341-H302-H317;
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 3.367
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number GF9800000
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H350-H360-H341-H302-H317;
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.