missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cobalt(II) chloride, Light blue beads, 99.9% (Metals basis), Alfa Aesar™
Sensitivity: Hygroscopic; 3.367. Decomposes at 400°C on long heating in air
Manufacturer: Alfa Aesar 1366606
Specifications
Cobalt(II) chloride | |
7646-79-9 | |
Cl2Co | |
cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii chloride, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride | |
Beads | |
Blue |
Ultra dry | |
99.9% (Metals basis) | |
MFCD00010938 | |
129.84 | |
Odorless | |
5g |
Chemical Identifiers
7646-79-9 | |
Cl2Co | |
MFCD00010938 | |
cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii chloride, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride |
Safety and Handling
H334-H350-H360-H317-H341-H400-H410-H302; May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. May cause an allergic skin reaction. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed.
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 231-589-4