missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) chloride hexahydrate, Puratronic™, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 1069218

 View more versions of this product

Catalog No. AA1069218



Specifications

Specifications

Cobalt(II) chloride hexahydrate
(metals basis)
MFCD00149652
14,2437
Crystalline Aggregates
Puratronic™
Odorless
Purple-Red
4.9
7791-13-1
CoCl2·6H2O
UN3077
237.93 (129.84 Anhydrous)
99.998%
1.924g/mL
Hygroscopic
87°C
50g

Chemical Identifiers

7791-13-1
MFCD00149652
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : GG0200000

TSCA : TSCA

Documents