Learn More
Cobalt(II) chloride hexahydrate, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 192862500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cobalt(II) chloride hexahydrate | |
7791-13-1 | |
Cl2CoH12O6 | |
MFCD00149652 | |
15,2421 | |
GFHNAMRJFCEERV-UHFFFAOYSA-L | |
λ²-cobalt(2+) hexahydrate dichloride | |
237.93 | |
Pure |
86.0°C | |
250 g | |
CoCl2·6H2O | |
01,155; 06,127; 11,479 | |
Solubility in water: 970g/L (20°C). | |
O.O.O.O.O.O.[Cl-].[Cl-].[Co++] | |
237.92 | |
98.0 to 102.0% (Complexometry) |
Chemical Identifiers
7791-13-1 | |
237.92 | |
GFHNAMRJFCEERV-UHFFFAOYSA-L | |
O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
Cl2CoH12O6 | |
MFCD00149652 | |
λ²-cobalt(2+) hexahydrate dichloride |
Safety and Handling
GHS H Statement
May damage fertility.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing genetic defects.
May cause cancer by inhalation.
May cause an allergic skin reaction.
May cause allergy or ast
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:
GHS Signal Word: Danger
RUO – Research Use Only