missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) chloride, 0.5M aq. soln.

$72.93 - $72.93

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2Co
MDL Number MFCD00149652
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63246AE View Documents Alfa Aesar™
J63246AE
100mL Each for $72.93
 
Specifications

Chemical Identifiers

7791-13-1
Cl2Co
MFCD00149652

Specifications

7791-13-1
CoCl2
UN3082
Liquid
Colorless
Cobalt(II) chloride
Cl2Co
MFCD00149652
129.84
0.5M aq. soln.
100mL
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : TSCA