Please login to your online account to display your discounted pricing

Cobalt(II) carbonate hydrate, ACROS Organics™


Quantity 50g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  192100500

 View more versions of this product

Catalog No. AC192100500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥45.5% (Cobalt) (Complexometry)
Physical Form Mass, Crystals, Crystalline Powder and/or Chunks
Formula Weight 118.94
Quantity 50g
Color White to yellow
EINECS Number 208-169-4
RTECS Number FF9450050
TSCA TSCA
Packaging Plastic bottle
Copper (Cu) 100ppm max.
Sodium (Na) 6000ppm max.
Health Hazard 3 GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Obtain special instructions before use. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS P Statement
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer by inhalation.
May cause an allergic skin reaction.
Harmful if swallowed.
May damage fertility.
Molecular Formula CCoO3·xH2O
MDL Number MFCD00149654
CAS 57454-67-8
Chemical Name or Material Cobalt(II) carbonate hydrate
Linear Formula CoCO3·xH2O
Chlorine (Cl) 100ppm max.
Iron (Fe) 100ppm max.
Sulfate 2000ppm max.
Cadmium (Cd) 50ppm max.
Insoluble Matter 0.05% max. (in HCl)
Health Hazard 1 Danger

Specifications

Chemical Name or Material Cobalt(II) carbonate hydrate
Assay ≥45.5% (Cobalt) (Complexometry)
Molecular Formula CCoO3·xH2O
Linear Formula CoCO3·xH2O
Formula Weight 118.94
MDL Number MFCD00149654
Accessories

Danger

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Obtain special instructions before use. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer by inhalation.
May cause an allergic skin reaction.
Harmful if swallowed.
May damage fertility.

SDS & Safety Documents