Please login to your online account to display your discounted pricing

Cobalt(II) bromide, ultra dry, 99.99% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Cobalt(II) bromide
CAS 7789-43-7
Physical Form Ultra Dry
Assay Percent Range 99.99% (Metal basis)
Molecular Formula CoBr2
Formula Weight 218.75
MDL Number MFCD00016017
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4031903

  SDS
alfa aesar
4031903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4031906

  SDS
alfa aesar
4031906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Ultra Dry
Formula Weight 218.75
Quantity 1g
Color Green
EINECS Number 232-166-7
Density 4.909g/cm3
Appearance Green (powder/flake/crystalline/beads, etc.)
MDL Number MFCD00016017
Health Hazard 1 H334-H315-H319-H317-H351-H302; May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. Suspected of causing cancer. Harmful if swallowed.
Molecular Formula CoBr2
Assay Percent Range 99.99% (Metal basis)
Chemical Name or Material Cobalt(II) bromide
CAS 7789-43-7
Safety and Handling

H334-H315-H319-H317-H351-H302; May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. Suspected of causing cancer. Harmful if swallowed.