Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) bromide, anhydrous, 97%

Chemical Identifiers

Chemical Name or Material Cobalt(II) bromide
CAS 7789-43-7
Name Note Anhydrous
Formula Weight 218.75
MDL Number MFCD00016017
Merck Index 14,2435
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2314918

  SDS
alfa aesar™
2314918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2314930

  SDS
alfa aesar™
2314930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 678°C (in N2)
Formula Weight 218.75
Sensitivity Hygroscopic
CAS 7789-43-7
TSCA TSCA
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number GF9595000
Quantity 50g
Chemical Name or Material Cobalt(II) bromide
Assay 97%
Physical Form Crystalline
Density 4.909g/mL
MDL Number MFCD00016017
Merck Index 14,2435
Name Note Anhydrous
Health Hazard 1 Danger
Solubility Information Soluble in water, methanol, ethanol, acetone, methyl acetate
Health Hazard 2 GHS H Statement
H334-H351-H302-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Color Green
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H351-H302-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.