missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) benzoate

Manufacturer:  Alfa Aesar™ 8738030

 View more versions of this product

Catalog No. AA8738030



Specifications

Specifications

Cobalt (II) benzoate
C14H10CoO4
MFCD00050769
C1=CC=C(C=C1)C(=O)[O-].C1=CC=C(C=C1)C(=O)[O-].[Co+2]
301.163
301.16
Gray
250g
Hygroscopic
932-69-4
Co(OOCC6H5)2
GAYAMOAYBXKUII-UHFFFAOYSA-L
cobalt(2+);dibenzoate
164760
Crystalline
114°C to 116°C
Odorless

Chemical Identifiers

932-69-4
301.163
GAYAMOAYBXKUII-UHFFFAOYSA-L
cobalt(2+);dibenzoate
C14H10CoO4
MFCD00050769
164760
C1=CC=C(C=C1)C(=O)[O-].C1=CC=C(C=C1)C(=O)[O-].[Co+2]
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents