Learn More
Cobalt(II) acetylacetonate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 210642500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cobalt(II) acetylacetonate | |
Authentic | |
Glass bottle | |
(CH3COCH=C(CH3)O)2Co | |
250g | |
17,87 | |
BKFAZDGHFACXKY-UHFFFAOYSA-N | |
257.14 |
14024-48-7 | |
98.5 to 101.5% (ex Co) | |
C10H14CoO4 | |
MFCD00000014 | |
01, 783 | |
Solubility in water: 5g/L (20°C). Other solubilities: 7g/kg alcohol (20°C),173g/kg benzene (20°C) | |
257.14 | |
99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Danger
EINECSNumber : 237-855-6
RUO â Research Use Only