missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt(II) acetate, Pale pink to purple powder, Alfa Aesar™

Manufacturer:  Alfa Aesar 2313814

 View more versions of this product

Catalog No. AA2313814



Specifications

Specifications

Cobalt(II) acetate
71-48-7
C4H6CoO4
QAHREYKOYSIQPH-UHFFFAOYSA-L
cobalt(2+);diacetate
6277
177.02
Odorless
Pink to Purple
Anhydrous, Co 32% min.
32% min. as Co
MFCD00008689
CC(=O)[O-].CC(=O)[O-].[Co+2]
177.021
CHEBI:85138
Powder
Anhydrous
25g

Chemical Identifiers

71-48-7
177.021
QAHREYKOYSIQPH-UHFFFAOYSA-L
CHEBI:85138
C4H6CoO4
MFCD00008689
6277
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

H334-H350-H360-H317-H341-H400-H410; May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. May cause an allergic skin reaction. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 200-755-8

SDS Documents