Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) 2-methoxyethoxide, 5% w/v in 2-methoxyethanol, 99% (m.b.), packaged under Ar in resealable ChemSeal bottles

Chemical Identifiers

Chemical Name or Material Cobalt (II) 2-methoxyethoxide
CAS 142600-61-1
Molecular Formula C6H14CoO4
Linear Formula Co(OCH2CH2OCH3)2
Formula Weight 209.11
MDL Number MFCD00798554
UN Number UN3274
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42819AC

  SDS
alfa aesar™
42819AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42819AE

  SDS
alfa aesar™
42819AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 209.11
Chemical Name or Material Cobalt (II) 2-methoxyethoxide
Physical Form Liquid
Assay 99%
Assay Percent Notes (metals basis)
Packaging ChemSeal™ bottles
Molecular Formula C6H14CoO4
Linear Formula Co(OCH2CH2OCH3)2
MDL Number MFCD00798554
CAS 142600-61-1
UN Number UN3274
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Packaged under Argon
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
Concentration 5% w/v in 2-methoxyethanol
Sensitivity Air & Moisture Sensitive
Quantity 25mL

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

  • Packaged under Argon