missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) 2,4-pentanedionate

Manufacturer:  Alfa Aesar™ 1253718

 View more versions of this product

Catalog No. AA1253718



Specifications

Specifications

Cobalt(II) 2,4-pentanedionate
Hygroscopic
165°C to 170°C
14024-48-7
MFCD00000014
BKFAZDGHFACXKY-UHFFFAOYSA-N
257.14
Odorless
Blue-Purple
50g
C10H14CoO4
Cobalt(II) acetylacetonate; Bis(acetylacetonato)cobalt(II)
257.15
Crystalline

Chemical Identifiers

14024-48-7
257.15
BKFAZDGHFACXKY-UHFFFAOYSA-N
C10H14CoO4
MFCD00000014
Cobalt(II) acetylacetonate; Bis(acetylacetonato)cobalt(II)
Safety and Handling

Safety and Handling

GHS H Statement
H334-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents