missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cobalt ICP Standard, 10,000 ppm Co in 5% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PCO10KN500
Specifications
Cobalt ICP Standard | |
Natural Poly Bottle | |
84.0,12.69,0.85 | |
Pink | |
Co | |
GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
cobalt | |
104730 | |
1 mL = 10 mg Co |
7440-48-4,7697-37-2,7440-48-4 | |
500 mL | |
88.0,13.21,0.88 | |
10,000ppm Co | |
MFCD00010935 | |
[Co] | |
58.93 | |
CHEBI:27638 |
Chemical Identifiers
7440-48-4 | |
58.93 | |
GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
CHEBI:27638 | |
[Co] |
Co | |
MFCD00010935 | |
104730 | |
cobalt |