Please login to your online account to display your discounted pricing

Cobalt ICP Standard, 1mL = 10mg Co (10,000ppm Co), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Cobalt ICP Standard
CAS 7732-18-5
CAS Min % 84.0
CAS Max % 88.0
CAS 7697-37-2
CAS Min % 12.69
CAS Max % 13.21
CAS 7440-48-4
CAS Min % 0.85
CAS Max % 0.88
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PCO10KN-100

 
ricca chemical company
RIPCO10KN100
Poly natural 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PCO10KN-50

 
ricca chemical company
RIPCO10KN50
Poly natural 50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PCO10KN-500

 
ricca chemical company
RIPCO10KN500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear light pink
Physical Form Liquid
Quantity 100mL
Chemical Name or Material Cobalt ICP Standard
CAS 7732-18-5
CAS Max % 0.88
CAS Min % 84.0
CAS Max % 88.0
CAS 7697-37-2
Packaging Poly natural
CAS Min % 12.69
CAS Max % 13.21
Concentration or Composition Notes 10,000ppm Co
CAS 7440-48-4
Equivalence 1 mL = 10 mg Co
CAS Min % 0.85