Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt foil, 1.0mm (0.04 in.) thick, Puratronic™, 99.995% (metals basis)

Chemical Identifiers

Chemical Name or Material Cobalt foil
CAS 7440-48-4
Name Note 1.0mm (0.04 in.) thick
Purity Grade Puratronic™
MDL Number MFCD00010935
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11450FF

  SDS
alfa aesar™
11450FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11450FI

  SDS
alfa aesar™
11450FI
50x50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Cobalt foil
TSCA TSCA
Name Note 1.0mm (0.04 in.) thick
Purity Grade Puratronic™
Physical Form Foil
Assay 99.995%
Assay Percent Notes (metals basis)
MDL Number MFCD00010935
CAS 7440-48-4
Odor Odorless
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Weight ≈5.6g/25x25mm
Quantity 25x25mm
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.