missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt foil, 0.25mm (0.01 in.) thick, Puratronic™, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 11452GH

 View more versions of this product

Catalog No. AA11452GH



Specifications

Specifications

Cobalt foil
100 x 100mm
7440-48-4
Co
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
[Co]
Odorless
104730
Foil
Puratronic™
≈5.6g/50x50mm
0.25mm (0.01 in.) thick
(metals basis)
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
58.933
CHEBI:27638
99.995%

Chemical Identifiers

7440-48-4
58.933
GUTLYIVDDKVIGB-UHFFFAOYSA-N
104730
cobalt
Co
MFCD00010935
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
CHEBI:27638
[Co]
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents