Please login to your online account to display your discounted pricing

Cobalt Chloride, 6-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$352.71 - $3,049.28

Chemical Identifiers

Chemical Name or Material Cobalt Chloride
CAS 7791-13-1
Name Note 6-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CoCl2·6H2O
Formula Weight 237.93
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002626

 
avantor performance
167004
Poly Bottle 125g Each for $352.71

02002627

 
avantor performance
167005
Poly Bottle 2.5kg Each for $3,049.28

02002628

 
avantor performance
167001
Poly Bottle 500g Each for $726.03
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 237.93
Chemical Name or Material Cobalt Chloride
Name Note 6-Hydrate
Molecular Formula CoCl2·6H2O
Packaging Poly Bottle
Specific Gravity 1L = 1.92kg
CAS 7791-13-1
Quantity 125g