Please login to your online account to display your discounted pricing

Cobalt Chloride, 6-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  167005

 View more versions of this product

Catalog No. 02002627

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 237.93
Chemical Name or Material Cobalt Chloride
Specific Gravity 1L = 1.92kg
Molecular Formula CoCl2·6H2O
CAS 7791-13-1
Quantity 2.5kg
Name Note 6-Hydrate
Packaging Poly Bottle

Specifications

Chemical Name or Material Cobalt Chloride
CAS 7791-13-1
Name Note 6-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CoCl2·6H2O
Formula Weight 237.93