missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt Chloride, 1.2% (w/v) in 10% (v/v) Hydrochloric Acid, Ricca Chemical
$150.09 - $229.88
Chemical Identifiers
| CAS | 7791-13-1 |
|---|---|
| Molecular Formula | Cl2CoH12O6 |
| Molecular Weight (g/mol) | 237.92 |
| MDL Number | MFCD00149652 |
| InChI Key | GFHNAMRJFCEERV-UHFFFAOYSA-L |
| Synonym | Cobalt (II) Chloride, Cobalt muriate hexahydrate |
| PubChem CID | 24643 |
| ChEBI | CHEBI:53503 |
| IUPAC Name | λ2-cobalt(2+) hexahydrate dichloride |
| SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
Chemical Identifiers
| 7791-13-1 | |
| 237.92 | |
| GFHNAMRJFCEERV-UHFFFAOYSA-L | |
| 24643 | |
| λ2-cobalt(2+) hexahydrate dichloride |
| Cl2CoH12O6 | |
| MFCD00149652 | |
| Cobalt (II) Chloride, Cobalt muriate hexahydrate | |
| CHEBI:53503 | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
Specifications
| Pink | |
| 1.04,87.0,10.23 | |
| Cl2CoH12O6 | |
| Cobalt (II) Chloride, Cobalt muriate hexahydrate | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Co++] | |
| 237.92 | |
| CHEBI:53503 | |
| Laboratory | |
| Aqueous Solution with 10% (v/v) Hydrochloric Acid | |
| 500 mL | |
| 1.2% (w/v) |
| 7791-13-1,7732-18-5,7647-01-0 | |
| 1.08,90.0,10.65 | |
| MFCD00149652 | |
| GFHNAMRJFCEERV-UHFFFAOYSA-L | |
| λ2-cobalt(2+) hexahydrate dichloride | |
| 24643 | |
| 1.2% | |
| Natural Poly Bottle | |
| <1 | |
| Liquid | |
| Cobalt Chloride |