Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt boride, 99%

Chemical Identifiers

Chemical Name or Material Cobalt boride
CAS 12619-68-0
MDL Number MFCD00146483
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1311706

 
alfa aesar™
1311706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1311714

 
alfa aesar™
1311714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Cobalt boride
TSCA TSCA
Mesh Size −10 Mesh
Odor Odorless
Assay 99%
MDL Number MFCD00146483
CAS 12619-68-0
Color Dark gray
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.