missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt boride, 99%

Manufacturer:  Alfa Aesar™ 1311714

 View more versions of this product

Catalog No. AA1311714



Specifications

Specifications

Cobalt boride
B2Co3+6
boron;cobalt 2+
QSOMCIJHXYGMAO-UHFFFAOYSA-N
boron;cobalt(2+)
121233614
99%
Odorless
25g
12619-68-0
MFCD00146483
Insoluble in water.
[B].[B].[Co+2].[Co+2].[Co+2]
198.42
-10 Mesh
−10 Mesh
Gray

Chemical Identifiers

12619-68-0
198.42
QSOMCIJHXYGMAO-UHFFFAOYSA-N
121233614
[B].[B].[Co+2].[Co+2].[Co+2]
B2Co3+6
MFCD00146483
boron;cobalt 2+
boron;cobalt(2+)
Safety and Handling

Safety and Handling

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350

EINECSNumber : 235-722-7

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents