missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt aluminum oxide, Co 39-41%

Manufacturer:  Alfa Aesar™ 1768622

 View more versions of this product

Catalog No. AA1768622



Specifications

Specifications

Cobalt aluminum oxide
Al2Co2O5
cobalt aluminate, aluminum cobalt oxide, aluminum cobalt oxide al2coo4, cobalt aluminate 100g, dialuminium cobalt tetraoxide, dialuminum; cobalt 2+ ; oxygen 2-, dialuminium 3+ dicobalt 2+ pentakis oxide 2-, dialuminium 3+ ion bis ? 2-cobalt 2+ ion pentaoxidandiide
[O-2].[O-2].[O-2].[O-2].[O-2].[Al+3].[Al+3].[Co+2].[Co+2]
251.824
176.89
Co 39 to 41%
Odorless
12672-27-4
MFCD00016016
CRHLEZORXKQUEI-UHFFFAOYSA-N
dialuminum;cobalt(2+);oxygen(2-)
159384
Powder
−200 Mesh
100g

Chemical Identifiers

12672-27-4
251.824
CRHLEZORXKQUEI-UHFFFAOYSA-N
159384
[O-2].[O-2].[O-2].[O-2].[O-2].[Al+3].[Al+3].[Co+2].[Co+2]
Al2Co2O5
MFCD00016016
cobalt aluminate, aluminum cobalt oxide, aluminum cobalt oxide al2coo4, cobalt aluminate 100g, dialuminium cobalt tetraoxide, dialuminum; cobalt 2+ ; oxygen 2-, dialuminium 3+ dicobalt 2+ pentakis oxide 2-, dialuminium 3+ ion bis ? 2-cobalt 2+ ion pentaoxidandiide
dialuminum;cobalt(2+);oxygen(2-)
Safety and Handling

Safety and Handling

GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents