missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt, AAS standard solution, Specpure™, Co 1000μg/mL

Manufacturer:  Alfa Aesar™ 88060AP

 View more versions of this product

Catalog No. AA88060AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cobalt, AAS standard solution
Co in 5% HNO3
UN3264
Specpure™
Co 1000μg/mL
MFCD00010935
Liquid
500mL

Chemical Identifiers

MFCD00010935
Safety and Handling

Safety and Handling

GHS H Statement
H334-H314-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents