Please login to your online account to display your discounted pricing

Cobalt AA Standard, 1mL = 1mg Co (1,000ppm Co) in 3% HCl, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Cobalt AA Standard
CAS 7732-18-5
CAS Min % 95.0
CAS Max % 98.0
CAS 7647-01-0
CAS Min % 3.31
CAS Max % 3.45
CAS 7440-48-4
CAS Min % 0.08
CAS Max % 0.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

ACO1KH-100

 
ricca chemical company
RIACO1KH100
Poly natural 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACO1KH-500

 
ricca chemical company
RIACO1KH500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear pink
Quantity 100mL
Chemical Name or Material Cobalt AA Standard
CAS 7732-18-5
CAS Min % 95.0
Concentration or Composition Notes 1,000ppm Co
Equivalence 1 mL = 1 mg Co
CAS 7647-01-0
CAS Max % 3.45
CAS 7440-48-4
CAS Max % 98.0
CAS Max % 0.08
CAS Min % 3.31
CAS Min % 0.08
Packaging Poly natural