Please login to your online account to display your discounted pricing

Co 102862, Tocris

State-dependent Na+ channel blocker; anticonvulsant

$172.26 - $726.06

Chemical Identifiers

Chemical Name or Material Co 102862
Assay Percent Range >99%
Molecular Formula C14H12FN3O2
Formula Weight 273.26
Synonym 2-[[4-(4-Fluorophenoxy)phenyl]methylene]hydrazinecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

264250

 
r & d systems
RDS264250
50mg Each for $726.06

264210

 
r & d systems
RDS264210
10mg Each for $172.26
Description & Specifications

Specifications

Formula Weight 273.26
Quantity 50mg
Chemical Name or Material Co 102862
Synonym 2-[[4-(4-Fluorophenoxy)phenyl]methylene]hydrazinecarboxamide
Assay Percent Range >99%
CAS 181144-66-1
Molecular Formula C14H12FN3O2
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature