Please login to your online account to display your discounted pricing

Co 101244 hydrochloride, Tocris

Highly selective NR2B antagonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material Co 101244 hydrochloride
Assay Percent Range >99%
Molecular Formula C21H27NO3·HCl
Formula Weight 377.91
Synonym 1-[2-(4-Hydroxyphenoxy)ethyl]-4-[(4-methylphenyl)methyl]-4-piperidinol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

245610

 
r & d systems
245610
10mg Each for $214.12

245650

 
r & d systems
245650
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 377.91
Quantity 10mg
Chemical Name or Material Co 101244 hydrochloride
Synonym 1-[2-(4-Hydroxyphenoxy)ethyl]-4-[(4-methylphenyl)methyl]-4-piperidinol hydrochloride
Assay Percent Range >99%
CAS 193356-17-1
Molecular Formula C21H27NO3·HCl
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C