Please login to your online account to display your discounted pricing

CNQX disodium salt, Tocris


Potent AMPA/kainate antagonist; More water soluble form of CNQX

Quantity 10mg

Manufacturer: r & d systems  RDS104510

 View more versions of this product

Catalog No. 10-451-0

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 276.12
Quantity 10mg
Synonym 6-Cyano-7-nitroquinoxaline-2, 3-dione disodium
CAS 479347-85-8
Assay Percent Range >99%
Molecular Formula C9H2N4O4Na2
Chemical Name or Material CNQX disodium salt
Recommended Storage Desiccate at Room Temperature

Specifications

Chemical Name or Material CNQX disodium salt
Assay Percent Range >99%
Molecular Formula C9H2N4O4Na2
Formula Weight 276.12
Synonym 6-Cyano-7-nitroquinoxaline-2, 3-dione disodium
Accessories

Desiccate at Room Temperature