Please login to your online account to display your discounted pricing

CNQX disodium salt, Tocris


Potent AMPA/kainate antagonist; More water soluble form of CNQX

Quantity 1mg

Manufacturer: r & d systems  10451

 View more versions of this product

Catalog No. 10451

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 276.12
Quantity 1mg
Chemical Name or Material CNQX disodium salt
Assay Percent Range >99%
Molecular Formula C9H2N4O4Na2
CAS 479347-85-8
Synonym 6-Cyano-7-nitroquinoxaline-2, 3-dione disodium
Recommended Storage Desiccate at Room Temperature

Specifications

Chemical Name or Material CNQX disodium salt
Assay Percent Range >99%
Molecular Formula C9H2N4O4Na2
Formula Weight 276.12
Synonym 6-Cyano-7-nitroquinoxaline-2, 3-dione disodium
Accessories

Desiccate at Room Temperature

SDS & Safety Documents