Please login to your online account to display your discounted pricing

CNQX, ACROS Organics™


Quantity 25mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  329070250

 View more versions of this product

Catalog No. AC329070250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Color Off-white to yellow
Quantity 25mg
Chemical Name or Material CNQX
CAS 115066-14-3
Physical Form Off-white to yellow powder
Molecular Formula C9H4N4O4
Formula Weight 232.20g/mol
MDL Number MFCD00069232
Synonym 6-Cyano-7-nitroquinoxaline-2, 3-dione

Specifications

Chemical Name or Material CNQX
CAS 115066-14-3
Molecular Formula C9H4N4O4
Formula Weight 232.20g/mol
MDL Number MFCD00069232
Synonym 6-Cyano-7-nitroquinoxaline-2, 3-dione