Please login to your online account to display your discounted pricing

CNQX, ACROS Organics™

Chemical Identifiers

Chemical Name or Material CNQX
CAS 115066-14-3
Molecular Formula C9H4N4O4
Formula Weight 232.20g/mol
MDL Number MFCD00069232
Synonym 6-Cyano-7-nitroquinoxaline-2, 3-dione
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC329070250

  SDS
thermo fisher scientific
329070250
Glass bottle 25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC329070050

  SDS
fisher scientific acros
329070050
Glass bottle 5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color Off-white to yellow
Quantity 25mg
Chemical Name or Material CNQX
CAS 115066-14-3
Physical Form Off-white to yellow powder
Molecular Formula C9H4N4O4
Formula Weight 232.20g/mol
MDL Number MFCD00069232
Synonym 6-Cyano-7-nitroquinoxaline-2, 3-dione