Please login to your online account to display your discounted pricing

CMPDA, Tocris™

Positive allosteric modulator of GluR2 receptors

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material CMPDA
Assay Percent Range >98%
Molecular Formula C16H28N2O4S2
Formula Weight 376.53
Synonym N,N'-(1,4-Phenylenedi-2,1-ethanediyl)bis-2-propanesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

437310

 
r & d systems
RDS437310
10mg Each for $225.77

437350

 
r & d systems
RDS437350
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 376.53
Quantity 10mg
Chemical Name or Material CMPDA
Assay Percent Range >98%
Molecular Formula C16H28N2O4S2
CAS 380607-77-2
Synonym N,N'-(1,4-Phenylenedi-2,1-ethanediyl)bis-2-propanesulfonamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C