Please login to your online account to display your discounted pricing

Alfa Aesar™ CLP Protocol Test Kit for ICP Analysis, Specpure™

Chemical Identifiers

Chemical Name or Material CLP Protocol Test Kit for ICP Analysis
Purity Grade Specpure™
MDL Number MFCD00146473
UN Number UN3264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35406KS

  SDS
alfa aesar™
35406KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material CLP Protocol Test Kit for ICP Analysis
Quantity 1 each
TSCA TSCA
Purity Grade Specpure™
MDL Number MFCD00146473
UN Number UN3264
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.