Please login to your online account to display your discounted pricing

Alfa Aesar™ Cloxacillin sodium monohydrate, 95%

Chemical Identifiers

Chemical Name or Material Cloxacillin sodium monohydrate
CAS 7081-44-9
Formula Weight 475.88
MDL Number MFCD00150735
Merck Index 14,2419
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6650003

  SDS
alfa aesar™
J6650003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 475.88
Chemical Name or Material Cloxacillin sodium monohydrate
Assay 95%
Physical Form Powder
MDL Number MFCD00150735
CAS 7081-44-9
Merck Index 14,2419
Boiling Point 69°C
Solubility Information Soluble in water
Color White to off-white
Melting Point 170°C
Storage Note 1 2° to 8°C
Quantity 1g
RTECS Number XH8920000

  • Targets primarily gram positive bacteria, especially _x001A_-lactamase producing Staphylococci

Safety and Handling

2° to 8°C