Please login to your online account to display your discounted pricing

Cloud Chamber Needle Sources

Designed specifically for use in cloud chambers.

$128.70 - $128.70

Specifications

For Use With (Application) Cloud chamber
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S49993

 
fisher science education
NSPB210
Pb-210 Needle Each for $128.70
Description & Specifications

Specifications

For Use With (Application) Cloud chamber

  • Activity is deposited into the eye of a needle to produce a point source
  • Packaged in a test tube with the needle inserted on the inside of the stopper

  • Polonium Emitter 0.1 μCi has a half-life of 138 days
  • Strontium Emitter 0.1 μCi has a half-life of 28.5 years
  • Lead Emitter 0.1 μCi has a half-life of 20.4 years