Thermo Scientific™ Closures with Prefitted Septa for 13mm Crimp/Snap-Ring LoVials™
Thermo Scientific™ National Closures with Prefitted Septa are available in easy-to-apply Kim-Snap or standard aluminum variety to suit your application needs.
$136.51 - $136.51
Specifications
Vial Closure Size 13mm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Septum Type Price Quantity  
Catalog Number Mfr. No. Septum Type Price Quantity  
03-397-36 Thermo Scientific™
C401553
Red PTFE/White Silicone/Red PTFE Snap Cap Pack of 100 Each for $136.51
Case of 10 Pack for $1,365.10
Specifications
Description
Thermo Scientific™ National Closures with Prefitted Septa are available in easy-to-apply Kim-Snap or standard aluminum variety to suit your application needs.
  • Kim-Snap™ closures provide a tight seal without the need of a crimper
  • National Scientific aluminum crimp closures provide a secure, leak-resistant seal
  • Aluminum seals must be applied with a crimping tool
Description & Specifications
Specifications
13mm