missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Clostripain, Clostridium histolyticum

A protease that cleaves proteins on the carboxyl bond of arginine

$432.54 - $432.54

Chemical Identifiers

CAS 9028-00-6
MDL Number MFCD00130814
Synonym Endoproteinase-Arg-C; Clostridiopeptidase b
Chemical Name or Material Clostripain, Clostridium histolyticum
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61362MCR SDS Alfa Aesar™
J61362MCR
1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ61362MA SDS Alfa Aesar™
J61362MA
10mg Each for $432.54
Specifications Description & Specifications

Chemical Identifiers

9028-00-6
MFCD00130814
Endoproteinase-Arg-C; Clostridiopeptidase b
Clostripain, Clostridium histolyticum

Specifications

9028-00-6
MFCD00130814
White
Clostripain, Clostridium histolyticum
Powder
Endoproteinase-Arg-C; Clostridiopeptidase b
1mg