Please login to your online account to display your discounted pricing

Alfa Aesar™ Clopamide, 98%


Quantity 500mg

Manufacturer: alfa aesar™  J67183ME

 View more versions of this product

Catalog No. AAJ67183ME

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 345.85
Melting Point 248° to 249°C
Chemical Name or Material Clopamide
MDL Number MFCD00941495
Assay 98%
CAS 636-54-4
Synonym 4-Chloro-N-(2,6-dimethylpiperidinyl)-3-sulfamoylbenzamide
Physical Form Solid
Quantity 500mg
Color White
RTECS Number CV2338875

Specifications

Chemical Name or Material Clopamide
CAS 636-54-4
Formula Weight 345.85
MDL Number MFCD00941495
Synonym 4-Chloro-N-(2,6-dimethylpiperidinyl)-3-sulfamoylbenzamide