Please login to your online account to display your discounted pricing

Alfa Aesar™ Clopamide, 98%

Chemical Identifiers

Chemical Name or Material Clopamide
CAS 636-54-4
Formula Weight 345.85
MDL Number MFCD00941495
Synonym 4-Chloro-N-(2,6-dimethylpiperidinyl)-3-sulfamoylbenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67183ME

  SDS
alfa aesar™
J67183ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 248° to 249°C
Formula Weight 345.85
Chemical Name or Material Clopamide
MDL Number MFCD00941495
CAS 636-54-4
Assay 98%
Synonym 4-Chloro-N-(2,6-dimethylpiperidinyl)-3-sulfamoylbenzamide
Physical Form Solid
Quantity 500mg
Color White
RTECS Number CV2338875