missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Clofilium p-toluenesulfonate, 97+%

A potassium channel blocker

$171.18 - $748.08

Chemical Identifiers

CAS 92953-10-1
Molecular Formula C28H44ClNO3S
Molecular Weight (g/mol) 510.174
MDL Number MFCD00069233
InChI Key MOQZYUUHIWPDQC-UHFFFAOYSA-M
Synonym clofilium tosylate, unii-13zvf1051k, clofilium tosylateclofilium tosylate, 4-chloro-n,n-diethyl-n-heptylbenzenebutanaminium tosylate, dsstox_cid_25601, dsstox_rid_80994, n-4-4-chlorophenyl butyl-n,n-diethylheptan-1-aminium 4-methylbenzenesulfonate, dsstox_gsid_45601, benzenebutanaminium, 4-chloro-n,n-diethyl-n-heptyl-, salt with 4-methylbenzenesulfonicacid 1:1, gnf-pf-3889
PubChem CID 175533
IUPAC Name 4-(4-chlorophenyl)butyl-diethyl-heptylazanium;4-methylbenzenesulfonate
SMILES CCCCCCC[N+](CC)(CC)CCCCC1=CC=C(C=C1)Cl.CC1=CC=C(C=C1)S(=O)(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63552MB View Documents Alfa Aesar™
J63552MB
25mg Each for $171.18
 
AAJ63552MD View Documents Alfa Aesar™
J63552MD
250mg Each for $748.08
 
Description

Description

Applications
Clofilium p-toluenesulfonate is a K+ channel blocker; cardiac depressant; anti-arrhythmic.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

92953-10-1
510.174
MOQZYUUHIWPDQC-UHFFFAOYSA-M
175533
CCCCCCC[N+](CC)(CC)CCCCC1=CC=C(C=C1)Cl.CC1=CC=C(C=C1)S(=O)(=O)[O-]
C28H44ClNO3S
MFCD00069233
clofilium tosylate, unii-13zvf1051k, clofilium tosylateclofilium tosylate, 4-chloro-n,n-diethyl-n-heptylbenzenebutanaminium tosylate, dsstox_cid_25601, dsstox_rid_80994, n-4-4-chlorophenyl butyl-n,n-diethylheptan-1-aminium 4-methylbenzenesulfonate, dsstox_gsid_45601, benzenebutanaminium, 4-chloro-n,n-diethyl-n-heptyl-, salt with 4-methylbenzenesulfonicacid 1:1, gnf-pf-3889
4-(4-chlorophenyl)butyl-diethyl-heptylazanium;4-methylbenzenesulfonate

Specifications

White
25mg
C28H44ClNO3S
clofilium tosylate, unii-13zvf1051k, clofilium tosylateclofilium tosylate, 4-chloro-n,n-diethyl-n-heptylbenzenebutanaminium tosylate, dsstox_cid_25601, dsstox_rid_80994, n-4-4-chlorophenyl butyl-n,n-diethylheptan-1-aminium 4-methylbenzenesulfonate, dsstox_gsid_45601, benzenebutanaminium, 4-chloro-n,n-diethyl-n-heptyl-, salt with 4-methylbenzenesulfonicacid 1:1, gnf-pf-3889
CCCCCCC[N+](CC)(CC)CCCCC1=CC=C(C=C1)Cl.CC1=CC=C(C=C1)S(=O)(=O)[O-]
510.174
510.17
≥97%
98°C to 99°C
92953-10-1
MFCD00069233
MOQZYUUHIWPDQC-UHFFFAOYSA-M
4-(4-chlorophenyl)butyl-diethyl-heptylazanium;4-methylbenzenesulfonate
175533
Powder
Clofilium p-toluenesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CY9040870