missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Clofilium p-toluenesulfonate, 97+%

A potassium channel blocker

Manufacturer:  Alfa Aesar™ J63552MD

 View more versions of this product

Catalog No. AAJ63552MD



Specifications

Specifications

Clofilium p-toluenesulfonate
98°C to 99°C
92953-10-1
MFCD00069233
MOQZYUUHIWPDQC-UHFFFAOYSA-M
4-(4-chlorophenyl)butyl-diethyl-heptylazanium;4-methylbenzenesulfonate
175533
Powder
White
250mg
C28H44ClNO3S
4-chloro-n,n-diethyl-n-heptylbenzenebutanaminium tosylate
CCCCCCC[N+](CC)(CC)CCCCC1=CC=C(C=C1)Cl.CC1=CC=C(C=C1)S(=O)(=O)[O-]
510.174
510.17
≥97%

Chemical Identifiers

92953-10-1
510.174
MOQZYUUHIWPDQC-UHFFFAOYSA-M
175533
CCCCCCC[N+](CC)(CC)CCCCC1=CC=C(C=C1)Cl.CC1=CC=C(C=C1)S(=O)(=O)[O-]
C28H44ClNO3S
MFCD00069233
4-chloro-n,n-diethyl-n-heptylbenzenebutanaminium tosylate
4-(4-chlorophenyl)butyl-diethyl-heptylazanium;4-methylbenzenesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CY9040870

Documents