Please login to your online account to display your discounted pricing

Clofarabine, Tocris

Deoxycytidine kinase (dCK) substrate

$257.85 - $257.85

Chemical Identifiers

Chemical Name or Material Clofarabine
Assay Percent Range >99%
Molecular Formula C10H11ClFN5O3
Formula Weight 303.68
Synonym 2-Chloro-9-(2-deoxy-2-fluoro-β-D-arabinofuranosyl)-9H-purin-6-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

26-001-0

 
r & d systems
260010
10mg Each for $257.85
Description & Specifications

Specifications

Formula Weight 303.68
Quantity 10mg
Molecular Formula C10H11ClFN5O3
Chemical Name or Material Clofarabine
Assay Percent Range >99%
CAS 123318-82-1
Synonym 2-Chloro-9-(2-deoxy-2-fluoro-β-D-arabinofuranosyl)-9H-purin-6-amine
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C