Invitrogen™ CLK3 Recombinant Human Protein DFS Item
CLK3 is a dual-specificity protein kinase.
Manufacturer:  Invitrogen™ LSPV3985
 View more versions of this product
Catalog No. PV3985
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
CLK3 can interact with and cause the nuclear redistribution of SR proteins.
Specifications
Specifications
NP_003983, T123S, G133S
Frozen
86kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
CLK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
CLK3
Baculovirus
Human
GST Tag
Documents