Invitrogen™ CLK2 Recombinant Human Protein DFS Item
CLK2 is a serine/threonine kinase that phosphorylates serine/arginine-rich proteins as related to controlling RNA splicing.
Manufacturer:  Invitrogen™ LSPV4279
 View more versions of this product
Catalog No. PV4279
Description
Description
CLK2 is thought to link cell cycle progression, apoptosis, and telomere length regulation.
Specifications
Specifications
NP_003983
Frozen
70.3kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
CLK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
1mg
RUO - research use only
CLK2
Baculovirus
Human
GST Tag
Documents